Anti-ADAM5 Rabbit Polyclonal Antibody
Catalog # PRSI22-049
Supplier: ProSci Inc.
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:ADAM metallopeptidase domain 5 (pseudogene)
- Antigen-Symbol:ADAM5
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Human
- Western blot:Yes
- Form:Liquid
- Gen-ID:255926
- Antigen-Synonyme:a disintegrin and metalloproteinase domain 5|TMDCII|ADAM5P|tMDC II
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant protein of human ADAM5.
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Specifications
About this item
Type: Primary
Antigen: ADAM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human