Anti-MGP Rabbit Polyclonal Antibody
Catalog # PRSI19-662
Supplier: ProSci Inc.
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Matrix Gla protein
- Antigen-Symbol:MGP
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:4256
- Antigen-Synonyme:MGLAP|MGP|GIG36|NTI
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 20-103 of human MGP (NP_000891.2).
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Specifications
About this item
Type: Primary
Antigen: MGP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat