Technische Daten
- Antikörper-Typ:Primär
- Antigen-Name:interferon, gamma
- Antigen-Symbol:IFNG
- Klonalität:Monoklonal
- Klon:4S.B3
- Konjugation:FITC (Fluorescein Isothiocyanate)
- Durchflusszytometrie:Yes
- Wirt:Mouse
- Isotyp:IgG1 kappa
- Reaktivität:Human
- Kreuzadsorption:No
- Form:liquid
- Gen-ID:P01579
- Antigen-Synonyme:IFNG
- Lagerungspuffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Lagertemperatur:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Transporttemperatur:4 °C
- Reinigung:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- VE:0,05 mg
Technische Daten
Über diesen Artikel
The 4S.B3 is a neutralizing antibody that binds with the human Interferon-gamma (IFN-gamma) protein, a 15 to 17 kDa cytokine with significant antibacterial, antiviral, and antitumoral properties. When secreted by natural killer cells and by natural killer T lymphocytes, it regulates the immune response and supports adaptive immunity when produced by Th1 or CD8+ T lymphocytes. IFN-gamma plays an important role in the activation, the growth, and the differentiation of the macrophages, B and T lymphocytes, and natural killer cells. It interacts synergically with other cytokines, such as TNF-alpha, to inhibit proliferation of normal and transformed cells. IFN- gamma is the primary cytokine that defines Th-1 cells.The biological activity of IFN-gamma is not affected by glycosylation
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: 4S.B3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human