Anti-Cd8b1 Rat Monoclonal Antibody [clone: H35-17.2] (PE (Phycoerythrin))
PRSI76-718
- Antikörper-Typ:Primär
- Antigen-Name:CD8 antigen, beta chain 1
- Antigen-Symbol:Cd8b1
- Klonalität:Monoklonal
- Klon:H35-17.2
- Konjugation:PE (Phycoerythrin)
- Durchflusszytometrie:Yes
- Wirt:Rat
- Isotyp:IgG2b kappa
- Kreuzadsorption:No
- Form:liquid
- Gen-ID:P10300
- Antigen-Synonyme:Cd8b1
- Lagerungspuffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Lagertemperatur:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Transporttemperatur:4 °C
- Reinigung:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- VE:0,1 mg
The H35-17.2 monoclonal antibody specifically reacts the mouse cd8 beta, a 32 kDa chain also known as Ly-3 and Lyt-3. CD8 is a T cell surface antigen co-receptor that interacts with MHC class I molecules on antigen presenting cells. It is expressed as a heterodimer with the alpha cd8 chain. The H35-17.2 antibody blocks T cell-mediated cytolysis of allogeneic lymphoma cells.
Type: Primary
Antigen: Cd8b1
Clonality: Monoclonal
Clone: H35-17.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse