Order Entry
Switzerland
ContactUsLinkComponent
Anti-H1H3592P3 Recombinant Antibody
Anti-H1H3592P3 Recombinant Antibody
  PRSI10-776
New Product
Anti-H1H3592P3 Recombinant Antibody
  PRSI10-776
New Product

 

  • Antikörper-Typ:
    Primär
  • Klonalität:
    Recombinant
  • Konjugation:
    Unconjugated
  • Wirt:
  • Isotyp:
    IgG1 kappa
  • Reaktivität:
  • Environmentally Preferable:
  • Antigen-Synonyme:
    ELT1-01|NEOD-001|2A4
  • Modification:
    None
  • Lagerungspuffer:
    PBS buffer pH 7,5
  • Lagertemperatur:
    Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2…8 °C for 1 - 2 weeks. Store at –20 °C for 12 months. Store at –80 °C for long term storage.
  • Konzentration:
    Batch dependent
  • Transporttemperatur:
    20
  • Immunogen:
    Chimeric,Humanised/SAA1 [Homo sapiens]
  • Reinigung:
    >95%
  • Packung:
    Vial
  • VE:
    0,1 mg

 

 

Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity: