Anti-Timd4 Recombinant Antibody [Clone: 5G3]
Catalog # PRSI24-617
Supplier: ProSci Inc.
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Timd4
- Antigen-Symbol:Timd4
- Klonalität:Recombinant
- Klon:5G3
- Konjugation:Unconjugated
- ELISA:Yes
- Durchflusszytometrie:Yes
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG kappa
- Reaktivität:Mouse
- Western blot:Yes
- Kreuzadsorption:No
- Form:Liquid
- Gen-ID:276891
- Antigen-Synonyme:Spleen|TIMD4|T-cell immunoglobulin and mucin domain containing 4|knockout of lymphotoxin protein|TIMD-4|T-cell membrane protein 4|T-cell immunoglobulin and mucin domain-containing protein 4|SMUCKLER|TIM4|mucin-containing|TIM-4|T-cell immunoglobulin mucin receptor 4
- Modification:Unmodified
- Lagerungspuffer:PBS with 0,02% Proclin 300
- Lagertemperatur:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Transporttemperatur:Shipped on blue ice
- Immunogen:This antibody was raised by immunising female Lewis rats (Harlan Sprague-Dawley) with Tim-4:Ig fusion protein in complete Freund's adjuvan
- Validierte Anwendungen:Blocking,Therapeutics
- Packung:Vial
- VE:0,2 mg
Specifications
About this item
Type: Primary
Antigen: Timd4
Clonality: Recombinant
Clone: 5G3
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Mouse