Anti-Havcr1 Recombinant Antibody [Clone: 3B3]
Catalog # PRSI24-599
Supplier: ProSci Inc.
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:hepatitis A virus cellular receptor 1
- Antigen-Symbol:Havcr1
- Klonalität:Recombinant
- Klon:3B3
- Konjugation:Unconjugated
- Durchflusszytometrie:Yes
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG kappa
- Reaktivität:Mouse
- Kreuzadsorption:No
- Form:Liquid
- Gen-ID:171283
- Antigen-Synonyme:TIM1|TIM-1|KIM-1|CD365|T-cell immunoglobulin mucin receptor 1|T cell immunoglobulin and mucin domain-containing protein 1|HAVCR1|T-cell immunoglobulin and mucin domain containing 1|Kidney injury molecule 1|HAVcr-1|Hepatitis A virus cellular receptor 1 homolog|T cell membrane protein 1|TIMD-1
- Modification:Unmodified
- Lagerungspuffer:PBS with 0,02% Proclin 300
- Lagertemperatur:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Transporttemperatur:Shipped on blue ice
- Immunogen:This antibody was raised by immunising female Lewis strain rats (Harlan Sprague-Dawley) subcutaneously with mouse TIM-1–Ig in complete Freund's adjuvant (CFA), followed by multiple 'boosting' with mouse TIM-1–Ig in PB
- Validierte Anwendungen:Agonism
- Packung:Vial
- VE:0,2 mg
Specifications
About this item
Type: Primary
Antigen: hepatitis A virus cellular receptor 1
Clonality: Recombinant
Clone: 3B3
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Mouse