Anti-HVEM Recombinant Antibody [Clone: HMHV-1B18]
Catalog # PRSI24-587
Supplier: ProSci Inc.
Specifications
- Antikörper-Typ:Primär
- Klonalität:Recombinant
- Klon:HMHV-1B18
- Konjugation:Unconjugated
- Durchflusszytometrie:Yes
- Immunpräzipitation:Yes
- Isotyp:IgG kappa
- Reaktivität:Mouse
- Western blot:Yes
- Kreuzadsorption:No
- Form:Liquid
- Antigen-Synonyme:Herpes virus Entry Mediator A|TNF Receptor-like 2|HVEA|Herpes virus Entry Mediator|CD40-Like Protein|Tumor Necrosis Factor Receptor-Like 2|CD270|ATAR|TNFRSF14|TR2|Tumor Necrosis Factor Receptor-Like Gene2|LIGHTR|Tumor Necrosis Factor Receptor Superfamily Member 14
- Modification:Unmodified
- Lagerungspuffer:PBS with 0,02% Proclin 300
- Lagertemperatur:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Transporttemperatur:Shipped on blue ice
- Immunogen:This antibody was raised by immunising Armenian hamsters with mouse HVEM:Fc fusion protei
- Validierte Anwendungen:Blocking,Functional Assays
- Packung:Vial
- VE:0,2 mg
Specifications
About this item
Type: Primary
Antigen: HVEM
Clonality: Recombinant
Clone: HMHV-1B18
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Mouse