Lomustin ≥97% (durch HPLC)
ENZOALX400038M050
New Product
:
- VE:50 mg
- Seq:2370
- Environmentally Preferable:
- Purity:≥97% (HPLC)
- Identity:Determined by IR
: 1-(2-Chloroethyl)-3-cyclohexyl-1-nitrosourea, CCNU
:
:233.70 g/mol
:235-859-2
:GHS06
:GHS08
:Kühlschrank
:13010-47-4
:C₉H₁₆ClN₃O₂
:MFCD00012392