96-Well Autosampler Vial Plate, with Inserts
Katalognr. MICS95096-710-D
Anbieter: MICROSOLV
Technische Daten
- Bezeichnung:96 well plate with 1 ml glass inserts
- VE:250
Technische Daten
Über diesen Artikel
96 well plates with glass inserts.
- High grade, pure silicone rubber with PTFE coating
- Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass
- Chemically inert
The inserts are removable and the plates can be reused.