Hei-TORQUE Overhead Stirrers, Gold 2 Package
Catalog # HEID501-62029-04
Supplier: Heidolph Instruments GmbH & Co.KG
Specifications
- Bezeichnung:Hei-TORQUE Gold 2 package, CH-plug
- Seq:03
- Environmentally Preferable:
- VE:1
Specifications
About this item
In order to provide the perfect end-to-end solution for powerful stirring and the simplest operation in the laboratory, we have added packages to the Hei-TORQUE product group. Each package contains a telescopic stand and a suitable clamp for easy use on lab tables.
Delivery: Hei-TORQUE Ultimate 200 comes with telescope stand and clamp.