Human Sperm Antibody ELISA Kit
Catalog # AVIVOKCD01286
Anbieter: Aviva Systems Biology
CAS-Nummer:
Technische Daten
- VE:1 KIT
- Testtyp:Capture
- Probentyp:Serum, plasma and other biological fluids
- Erkennungsmethode:Colorimetric
- Lagertemperatur:2 to 8 °C or −20°C
- Anwendung:ELISA-Comp
- Empfindlichkeit:1,17 ng/ml