Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Versiegeln
Dichtringe
KF Outer Centering Ring with Viton O-Ring, Aluminium
KF Outer Centering Ring with Viton O-Ring, Aluminium
ILMV760531
:
Welch by Gardner Denver
KF Outer Centering Ring with Viton O-Ring, Aluminium
ILMV760531
:
Welch by Gardner Denver
Für:
Glass-to-metal flange connections
Bezeichnung:
KF outer centering ring with Viton O-ring
Material:
Aluminium
VE:
1