1-Ethyl-3-methylimidazoliumdicyanamid
Catalog # ROTH2053.1
Supplier: Roth Carl
CAS Number:
Specifications
- Packung:Glasflasche für Feststoffe
- VE:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: Basionics VS 03, EMIM DCA
:
:177.21 g/mol
CAS Number:370865-89-7
:MFCD28145696
Formula:C₈H₁₁N₅
MDL Number:MFCD08276373