Hydrometer for Lime Milk
HECH46199010
New Product
- Bezeichnung:Hydrometer for lime milk without thermometer
- Länge:~29 cm
- Referenztemperatur:20 °C
- VE:1
- Messbereich:0 - 340: 5,0 g/ml
Hydrometer for lime milk without thermometer.
Hydrometer for lime milk without thermometer.