Phthalaldehyd
Glass bottle for solids, 25 g
Katalognr. ROTH5980.1
Marketsource
Anbieter: Roth Carl
Direkt zu
Product Details & Documents
Synonyme: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
- CAS Nummer:643-79-8
- Formel:C₈H₆O₂
- MDL-Nummer:MFCD00003335
- Molekulargewicht:134.13 g/mol
- Siedepunkt:266 °C (1013 hPa)
- Schmelzender Platingehalt:54…56 °C
- Dichte:1,13 g/cm³ (20 °C)
- Blitz Pt:132 °C
- EINECS:211-402-2
- UN:2923
- ADR:8,II
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:25 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



