Order Entry
Switzerland
ContactUsLinkComponent
 
Irritant
2-Methyl-4-nitroanisol
Catalog # MOLE75004060
Supplier:  Molekula
CAS Number:  
Irritant
2-Methyl-4-nitroanisol
Catalog # MOLE75004060
Supplier:  Molekula
CAS Number:  

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :167.16 g/mol
Boiling Pt:311 °C (1013 hPa)
Melting Pt:96 °C
Storage Temperature:Raumtemperatur
EINECS:202-763-7
UN:2446
ADR:6.1,III
 :GHS07
CAS Number:50741-92-9
 :256-748-5
Formula:C₈H₉NO₃
MDL Number:MFCD07787583