AuftragsverfolgungKontakt
NucSpot®
NucSpot®
Supplier:   Biotium
NucSpot®
Supplier:  Biotium
Certificates
2 Options Available Below
Product Details & Documents
NucSpot® nuclear stains are cell membrane-impermeant, nuclear-specific counterstains available in a variety of colors from green to near-infrared (near-IR).
  • Nuclear-specific counterstains for fixed cells
  • Selectively stains dead cells in live culture
  • No wash required
  • Options for cell cycle profiling by flow cytometry in fixed/permeabilised cells
  • Color options for FITC, Cy®3, PE, Texas Red®, Cy®5, APC, Cy®5,5, and Cy®7 channels
NucSpot® stains have minimal fluorescence until they bind to DNA and can be used for no-wash nuclear staining. Unlike other nucleic acid dyes such as propidium iodide (PI), TOTO®, TO-PRO®, and similar dyes that stain both the nucleus and cytoplasm, NucSpot® nuclear stains selectively stain the nucleus in fixed and permeabilised cells without the need for RNase treatment.
NucSpot® nuclear stains also can be used for selective staining of dead cells in unfixed cell cultures for analysis by flow cytometry or fluorescence imaging. Several of the dyes can be continuously incubated with cells for multi-day imaging. NucSpot® 470 and NucSpot® far-red can be used for DNA content analysis of cell cycle by flow cytometry in fixed and permeabilised cells without requiring RNase treatment, unlike propidium iodide.
Product Family Options
Product Information
  • VE
    Availability
    Price
  • 20 µl
    Log in for availability
    Log in for availability
    1 * 20 µl
    1 * 20 µl - 167.00 SFr.
    167.00 SFr.
    1 * 20 µl
       167.00 SFr.
  • 100 µl
    Log in for availability
    Log in for availability
    1 * 100 µl
    1 * 100 µl - 742.00 SFr.
    742.00 SFr.
    1 * 100 µl
       742.00 SFr.