AuftragsverfolgungKontakt
iFluor® 790, Säure
iFluor® 790, Säure
5 mg
Catalog # AATB1360
Supplier:   AAT Bioquest
iFluor® 790, Säure
Catalog # AATB1360
Supplier:  AAT Bioquest
CAS Number:  
iFluor® 790, Säure
iFluor® 790, Säure
5 mg
CAS Number:  
Price
315.00 SFr.
Log in for availability
1 * 5 mg
Product Details & Documents
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
  • Bright fluorescence
  • High photostability
  • Low quenching on proteins
  • Useful for in-vivo imaging
Caution: For research use only.
  • Storage Temperature:Gefrierschrank
Specifications
Specifications
  • VE:
    5 mg