ContactUsLinkComponent
ReadiUse™ Mammalian Cell Lysis Buffer *5X*
Catalog # AATB20012
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiUse™ Mammalian Cell Lysis Buffer *5X*
Catalog # AATB20012
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • VE:
    10 mL
Specifications
About this item
All our ReadiUse™ reagents require minimal hands-on time.
  • 5x concentrated
This mammalian cell lysis buffer just require a simple 5-fold dilution..