MitoLite™ Red CMXRos
MitoLite™ red CMXRos, 500 µG
AATB22698
MitoLite™ red CMXRos is chemically same to the MitoTracker™ Red CMXRos (ThermoFisher).
- Mitochondria-Specific Dye
- Cell-Permeable
- Robust
Caution: For research use only.
- VE:500 µG
- Bezeichnung:MitoLite™ red CMXRos
- Cat. No.:AATB22698
- Purity:90%