Amplite® Fluorimetric Monoamine Oxidase Assay Kit *Red Fluorescence*
Katalognr. AATB11303
Anbieter: AAT BIOQUEST
CAS-Nummer:
Technische Daten
- VE:200 Tests
- Bezeichnung:Amplite™ fluorimetric monoamine oxidase assay kit *red fluorescence*
Technische Daten
Über diesen Artikel
Monoamine oxidases (MAO) are a family of flavin-containing amine oxidoreductases that catalyse the oxidation of monoamines.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.