Cell Meter™ Intracellular NADH/NADPH Flow Cytometric Analysis Kit *Deep Red Fluorescence*
Catalog # AATB15296
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- VE:100 Tests
- Bezeichnung:Cell Meter™ intracellular NADH/NADPH Flow cytometric analysis kit *deep red fluorescence*
Specifications
About this item
The detection of intracellular dihydronicotinamide adenine dinucleotide NADH and its phosphate ester NADPH is important for disease diagnostics and drug discovery.
- High sensitivity
- Bright fluorescence
- Suitable for live cell imaging
Caution: For research use only.