Amplite® Fluorimetric Glutathione Assay Kit *Green Fluorescence*
Catalog # AATB10055
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- VE:200 Tests
- Bezeichnung:Amplite™ fluorimetric glutathione assay kit *green fluorescence*
- Tests pro Kit:200 Tests
Specifications
About this item
The monitoring of reduced and oxidised glutathione (GSH) in biological samples is essential for evaluating the redox and detoxification status of cells and tissues in relation to the protective role of glutathione against oxidative and free-radical-mediated cell injury.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.