Amplite® Fluorimetric Acetylcholine Assay Kit *Red Fluorescence*
Catalog # AATB11403
Supplier: AAT Bioquest
CAS Number:
Specifications
- VE:200 Tests
- Bezeichnung:Amplite™ fluorimetric acetylcholine assay kit *red fluorescence*
Specifications
About this item
Acetylcholine and its metabolites are needed for three main physiological purposes: structural integrity and signaling roles for cell membranes, cholinergic neurotransmission (acetylcholine synthesis), and as a major source for methyl groups via its metabolite, trimethylglycine (betaine) that participates in the S-adenosylmethionine synthesis pathways.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.