ContactUsLinkComponent
Helixyte™ Green Fluorimetric ssDNA Quantitation Kit
Catalog # AATB17623
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Helixyte™ Green Fluorimetric ssDNA Quantitation Kit
Catalog # AATB17623
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • VE:
    200 Tests
  • Bezeichnung:
    Designed to measure single-stranded DNA in an easy and accurate format
Specifications
About this item
Helixyte™ Green fluorimetric ssDNA quantitation kit is optimised for quantifying dsDNA with a fluorescence microplate reader.
  • Optimized for microplate readers
  • High sensitivity
  • Low interference from common contaminants
The kit has all the essential reagents, including Helixyte™ Green ssDNA reagent, dilution buffer and prediluted DNA standards. Helixyte™ Green ssDNA reagent is a sensitive fluorescent nucleic acid probe for quantifying oligonucleotides and single-stranded DNA (ssDNA) in solution. It enables researchers to quantify as little as 100 pg/ml oligonucleotide or ssDNA. This sensitivity exceeds that achieved with absorbance methods by more than 10000-fold.
With this kit, as little as 1 ng/ml oligonucleotide or ssDNA might be detected.
Packaging: Plastic vial.
Caution: For research use only.