Cell Navigator® Lysosome Staining Kit *NIR Fluorescence*
Catalog # AATB22652
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- VE:500 Tests
- Bezeichnung:Cell Navigator™ Lysosome staining kit *NIR fluorescence*
- Tests pro Kit:500 Tests
Specifications
About this item
Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.
- Cell-permeant staining
- Improved signal
- Suitable for all major fluorescence platforms
Caution: For research use only.