Cell Meter™ Fixable Viability Dye, BX520
Cell Meter™ BX520 fixable cell stain, 200 Tests, 200 Tests
Katalognr. AATB22510
Anbieter: AAT Bioquest
Direkt zu
Product Details & Documents
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
- Cell-Impermeable
- Amine- And Thiol-Reactive
- High Stability
- Live/Dead Cell Labeling Dye
- Fixable
Achtung! Bitte beachten: For research use only.
Technische Daten
Technische Daten
- VE:200 Tests
- Bezeichnung:Cell Meter™ BX520 fixable cell stain
- Tests pro Kit:200 Tests