Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
AMCA Azide ≥95%

AMCA Azide ≥95%

1 mg
Catalog # AATB508
Supplier:   AAT Bioquest
AMCA Azide ≥95%
Catalog # AATB508
Supplier:  AAT Bioquest
CAS Number:  
AMCA Azide ≥95%
AMCA Azide ≥95%

1 mg

CAS Number:  
Price
SFr. 157.00
Log in for availability
1 * 1 mg
Product Details & Documents

AMCA is one of the brightest blue fluorescent labeling dyes.

  • Bright Fluorescence
  • Click Chemistry Conjugation Reagent

 : For research use only.

  • Molecular Weight:301.3 g/mol
Specifications

Specifications

  • VE:
    1 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.