Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Dihydroethidium ≥95%

Dihydroethidium ≥95%

Plastic bottle, 25 mg
Katalognr. AATB15200
Reizend
Anbieter:   AAT Bioquest
Dihydroethidium ≥95%
Katalognr. AATB15200
Anbieter:  AAT Bioquest
CAS Nummer:  104821-25-2
Dihydroethidium ≥95%
Dihydroethidium ≥95%

Plastic bottle, 25 mg

CAS Nummer:  104821-25-2
Preis
157.00 SFr.
Einloggen für Verfügbarkeit
1 * 25 mg
Product Details & Documents

Hydroethidine operates effectively as a probe for measurement of reactive oxygen species.

Synonyme:  3,8-Diamino-5,6-dihydro-5-ethyl-6-phenylphenanthridine, Hydroethidine, DHE, 5-Ethyl-6-phenyl-5,6-dihydrophenanthridine-3,8-diamine, 2,7-Diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine
  • CAS Nummer:104821-25-2
  • Formel:C₂₁H₂₁N₃
  • MDL-Nummer:MFCD00077335
  • Molekulargewicht:315.412 g/mol
  • ADR:5,II
Technische Daten

Technische Daten

  • Packung:
    Plastic bottle
  • VE:
    25 mg

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.