Order Entry
Switzerland
ContactUsLinkComponent
The DNP (Rat) IgG ELISA kit
The DNP (Rat) IgG ELISA kit
Catalog # ABNOKA6741.1KIT
Supplier:  Abnova
CAS Number:  
The DNP (Rat) IgG ELISA kit
Catalog # ABNOKA6741.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • VE:
    1 KIT
  • Reaktivität der primären Antikörper:
    Rat
  • Target protein:
    DNP

Specifications

About this item

The DNP (Rat) IgG ELISA Kit is based on a solid phase enzymelinked immunosorbent assay (ELISA). The assay uses DNP-BSA as the capture antigen (coated on the microtiter wells) and HRP conjugated anti-rat IgG antibodies for detection.