Anti-LMNB1 Rabbit Recombinant Antibody [clone: R05-8D6]
Catalog # ABNORAB02143.100UL
Supplier: Abnova
Specifications
- Antigen-Symbol:LMNB1
- Klonalität:Recombinant
- Klon:R05-8D6
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human LMNB1.
- VE:100 µl