Anti-HSP90AA1/HSP90AB1 Rabbit Recombinant Antibody [clone: R08-8A1]
ABNORAB02194.100UL
: Abnova
- Antigen-Symbol:HSP90AA1/HSP90AB1
- Klonalität:Recombinant
- Klon:R08-8A1
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human HSP90AA1/HSP90AB1.
- VE:100 µl