Anti-USP13 Rabbit Recombinant Antibody [clone: R08-8D3]
Catalog # ABNORAB01841.100UL
Supplier: Abnova
Specifications
- Antigen-Symbol:USP13
- Klonalität:Recombinant
- Klon:R08-8D3
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human USP13.
- VE:100 µl