Anti-Ntrk2 Rabbit Recombinant Antibody [clone: R03-4C7]
Catalog # ABNORAB01851.100UL
Supplier: Abnova
Specifications
- Antigen-Symbol:Ntrk2
- Klonalität:Recombinant
- Klon:R03-4C7
- Wirt:Rabbit
- Reaktivität:Mouse
- Immunogen:Recombinant rabbit monoclonal antibody raised against mouse Ntrk2.
- VE:100 µl