Anti-TRAF6 Rabbit Recombinant Antibody [clone: R05-2D6]
Catalog # ABNORAB01859.100UL
Supplier: Abnova
Specifications
- Antigen-Symbol:TRAF6
- Klonalität:Recombinant
- Klon:R05-2D6
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human TRAF6.
- VE:100 µl