Order Entry
Switzerland
ContactUsLinkComponent
Anti-CELF-2 Rabbit Recombinant Antibody [clone: R08-9E3]
Anti-CELF-2 Rabbit Recombinant Antibody [clone: R08-9E3]
Catalog # ABNORAB01506.100UL
Supplier:  Abnova
Anti-CELF-2 Rabbit Recombinant Antibody [clone: R08-9E3]
Catalog # ABNORAB01506.100UL
Supplier:  Abnova

Specifications

  • Antigen-Symbol:
    CELF-2
  • Klonalität:
    Recombinant
  • Klon:
    R08-9E3
  • Wirt:
    Rabbit
  • Reaktivität:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human CELF-2.
  • VE:
    100 µl

Specifications

About this item