Anti-ACO2 Rabbit Recombinant Antibody [clone: R07-7A1]
Catalog # ABNORAB01769.100UL
Supplier: Abnova
Specifications
- Antigen-Symbol:ACO2
- Klonalität:Recombinant
- Klon:R07-7A1
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human ACO2.
- VE:100 µl