Anti-KEAP1 Rabbit Recombinant Antibody [clone: R02-1H7]
Catalog # ABNORAB01248.100UL
Supplier: Abnova
Specifications
- Antigen-Symbol:KEAP1
- Klonalität:Recombinant
- Klon:R02-1H7
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human KEAP1.
- VE:100 µl