Anti-FUBP1 Rabbit Recombinant Antibody [clone: R01-5D4]
Catalog # ABNORAB01264.100UL
Supplier: Abnova
Specifications
- Antigen-Symbol:FUBP1
- Klonalität:Recombinant
- Klon:R01-5D4
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human FUBP1.
- VE:100 µl