Anti-TNF alpha Rabbit Recombinant Antibody [clone: D2E7 (ADALIMUMAB)]
Catalog # ABNORAB01186.200UG
Supplier: Abnova
Specifications
- Antigen-Symbol:TNF alpha
- Klonalität:Recombinant
- Klon:D2E7 (ADALIMUMAB)
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human TNF alpha.
- VE:200 µG