Ready-to-Load™ kits, in search of my father
Katalognr. EDVOS-49
Anbieter: EDVOTEK
Technische Daten
- Bezeichnung:In Search of My Father
- Environmentally Preferable:
- VE:1
Technische Daten
Über diesen Artikel
Solve the mystery of two boys separated from their parents a decade ago. Their biological mother is identified by mitrochondrial DNA and their biological father from chromosomal DNA.
- For 10 lab groups
- Complete in 45 minutes
Also required: Electrophoresis tank and power supply.
Delivery: Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.