ContactUsLinkComponent
Children's Milk Bottle, Wide Neck
  HECH51866010
Children's Milk Bottle, Wide Neck
  HECH51866010
  • Kapazität:
    250 ml
  • Farbe:
    Klar
  • Hals I-Ø:
    30 mm
  • VE:
    12
Children's milk bottle, wide mouth, with graduations from 20 to 200 ml.
  • Borosilicate glass 3.3
  • Graduated blue
  • Flat floor
  • Strong rolled edge