96 Well Plate, RCH2 PPBA
Catalog # TCAT30168995
Supplier: Tecan
Specifications
- Anz. Wells:96
- Bezeichnung:96-well RCH Atlas® plate
- Phase:RCH2
- Füllung:50 mg
- VE:1
Specifications
About this item
A special phenyl boronic acid chemistry on polymer base resin for better capacity and solvent use. PBA utilizes an unique covalent interaction ideal for catecholamine analysis.
- Cerex
- Reversed-phased
- Liquid handler compatible
Particle size: 60 µm