Bucket Liners
Irradiated liner for B99 bucket, 889×533,4 mm
Catalog # MICRBKL993521IRB
Supplier: MICRONOVA
Jump to:
Product Details & Documents
The unique LDPE/nylon laminate is a superb barrier material for lining totes to transport raw materials or to line buckets containing disinfectants or other cleaning solutions.
- Laminate is 75% LDPE, 25% nylon
- Excellent for disinfectants and cleaning solutions
- Ideal for product parts or raw material transfer
- Thickness of 3 mil
- Will hold up to autoclaving with liquid contents
- Available sterile (irradiated) or non sterile
Non sterile liners are bulk packed. Select sterile liners are available bulk packed.
Fits: B-99 series buckets.
Packaging: Flat packed, either individually double-bagged or in bulk packs. Packed in a case of 24 or case of 72 for bulk pack.
Specifications
- Bezeichnung:Irradiated liner for B99 bucket
- L×B:889×533,4 mm