Fit Test Solution
Catalog # MMMAFT-32
Supplier: 3M
Specifications
- Bezeichnung:Fit test solution, bitter, 55 ml bottle
- VE:6
Specifications
About this item
Replacement fit test solution for use with 3M™ qualitative fit test apparatus kits FT-10 (Sweet) and FT-30 (Bitter). The choice between the bitter and sweet test kit is a personal one, however a small percentage of the population are unable to detect saccharin, in which case the bitter test must be used.
- Fit test solution; bitter taste
- 55 ml bottle
- For occupational use only
Test kits and accessories designed to help organisations meet regulatory fit testing requirements for tight fitting respirators. This product can be used to validate face to respirator seal on any disposable respirator or half mask respirator.