Disconnectors, Tapered
Katalognr. KART7053700
Anbieter: KARTELL
Technische Daten
- Ext.Ø:14-15-16 mm
- Bezeichnung:Disconnectors
- VE:100
Technische Daten
Über diesen Artikel
These quick disconnector is useful for joining hoses of glass, rubber or plastic. Connectors fit tightly together. Replace expensive adapters.
Dimensions are shown as both nominal and exact as elasticity of hoses will influence the choice of size.
Accessories information: Tested according to reach regulation 1907/2006 Annex XVII.