Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-Eph receptor B1 Tyr968 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-10139R-A680
Supplier:  Bioss
Anti-Eph receptor B1 Tyr968 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-10139R-A680
Supplier:  Bioss

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Eph receptor B1 Tyr968
  • Klonalität:
    Polyklonal
  • Konjugation:
    Alexa Fluor® 680
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • ImmunFluoreszenz:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gen-ID:
    2047
  • Antigen-Synonyme:
    Eph receptor B1phospho Y968|EPH2|Tyrosine protein kinase receptor EPH 2|Elkh|NET|Eph tyrosine kinase 2|Ephrin type B receptor 1|Ephrin receptor Eph B1 precursor|Neuronally expressed EPH related tyrosine kinase|HEK 6|EphB1|EphB1phospho Y968Cek 6
  • Modification:
    Phosphorylation (Tyr968)
  • Lagerungspuffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    106 kDa
  • Lagertemperatur:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Konzentration:
    1 µg/µl
  • Transporttemperatur:
    4 °C
  • Immunogen:
    KLH conjugated synthetic phosphopeptide derived from human EphB1 around the phosphorylation site of Tyr968
  • Validierte Anwendungen:
    ICC
  • Reinigung:
    Purified by Protein A
  • Packung:
    Vial
  • VE:
    100 µl

Specifications

About this item

EphB1, previously known as Elk (eph like kinase), is a receptor tyrosine kinase of the highly tissue restricted family of eph proteins. EphB1 and other ephB family members are type 1 membrane spanning proteins, comprised of immunoglobulin, fibronectin type III, and cysteine rich subdomains in the ecto domain, and the single uninterrupted cytoplasmic tyrosine kinase domain upstream of a carboxyterminal sterile alpha motif (SAM) domain. EphB family proteins bind ephrins of the B class. EphB1 is expressed predominately in developing neural structures in embryos, and in vascular epithelium of kidney, and other tissues. Upon binding to alternatively oligomerized ephrin B1, EphB1 signals regulation of cell attachment and cell to cell assembly. Members of this protein family are implicated in neuronal and vascular cell targeting.

Type: Primary
Antigen: Eph receptor B1 Tyr968
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat