Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-CD33 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-1514R-CY3
 :  Bioss
Anti-CD33 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-1514R-CY3
 :  Bioss

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    CD33
  • Klonalität:
    Polyklonal
  • Konjugation:
    Cy3®
  • Durchflusszytometrie:
    Yes
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • ImmunFluoreszenz:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    101-200/334
  • Form:
    Liquid
  • Gen-ID:
    12489
  • Antigen-Synonyme:
    gp67|Siglec3|CD33|Siglec-3|Myeloid cell surface antigen CD33|Sialic acid-binding Ig-like lectin 3
  • Modification:
    Unmodified
  • Lagerungspuffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    40/37 kDa
  • Lagertemperatur:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Konzentration:
    1 µg/µl
  • Transporttemperatur:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from mouse CD33
  • Validierte Anwendungen:
    ICC
  • Reinigung:
    Purified by Protein A
  • Packung:
    Vial
  • VE:
    100 µl

 

 

Putative adhesion molecule of myelomonocytic-derived cells that mediates sialic-acid dependent binding to cells. Preferentially binds to alpha-2,6-linked sialic acid (By similarity). The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface.

Type: Primary
Antigen: CD33
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 101-200/334
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse