Anti-CD33 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-1514R-A680
: Bioss
- Antikörper-Typ:Primär
- Antigen-Name:CD33
- Klonalität:Polyklonal
- Konjugation:Alexa Fluor® 680
- Durchflusszytometrie:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Human,Mouse
- Western blot:Yes
- Epitop:101-200/334
- Form:Liquid
- Gen-ID:12489
- Antigen-Synonyme:gp67|Siglec3|CD33|Siglec-3|Myeloid cell surface antigen CD33|Sialic acid-binding Ig-like lectin 3
- Modification:Unmodified
- Lagerungspuffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:40/37 kDa
- Lagertemperatur:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Konzentration:1 µg/µl
- Transporttemperatur:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse CD33
- Validierte Anwendungen:ICC
- Reinigung:Purified by Protein A
- Packung:Vial
- VE:100 µl
Putative adhesion molecule of myelomonocytic-derived cells that mediates sialic-acid dependent binding to cells. Preferentially binds to alpha-2,6-linked sialic acid (By similarity). The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface.
Type: Primary
Antigen: CD33
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/334
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse